About this item
InVivoMab anti-mouse TIM-4, Clone: RMT4-53, Isotype: rat IgG2a, κ, Applications: in vivo TIM-4 blockade, in vitro TIM-4 blockade, Immunofluorescence
Type: Primary
Antigen: TIM-4
Clonality: Monoclonal
Clone: RMT4-53
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76556-052
- 76555-438
- 76554-652
- 76555-332
- 76557-120
- Antigen Symbol:
- TIM-4
- TIM-4
- TIM-4
- TIM-4
- TIM-4
- Antigen Name:
- TIM-4
- TIM-4
- TIM-4
- TIM-4
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- RMT4-53
- RMT4-53
- RMT4-53
- RMT4-53
- RMT4-53
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes