To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 20
- Antigen Symbol:USP20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 μL
- Environmentally Preferable:
- Epitope:814-844 aa
- Form:Liquid
- Gene ID:10868
- Modification:None
- Molecular Weight:102 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/ml
- Shipping Temperature:4°C
- Immunogen:This USP20 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 814-844 amino acids from the C-terminal region of human USP20.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76070-280
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: USP20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 814-844
Host: Rabbit
Isotype: Ig
Reactivity: Mouse