Order Entry
United States
ContactUsLinkComponent
 
Bilirubin (Total and Direct) Colorimetric Assay Kit, BioVision Inc.
Catalog # 76020-886
Supplier:  Avantor
CAS Number:  
undefined
Bilirubin (Total and Direct) Colorimetric Assay Kit, BioVision Inc.
Catalog # 76020-886
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    100 assays
  • Description:
    Bilirubin Colorimetric Assay Kit
  • Size:
    100 assays
  • Environmentally Preferable:
  • Brand Name:
    BioVision Inc.
  • Cat. No.:
    76020-886

Specifications

About this item

Allows measurement of both Total and Direct or Conjugated Bilirubin

  • Simple protocol, robust and reliable data
  • This assay allows detection of both Total and Direct or conjugated Bilirubin in serum samples.

Bilirubin, a degradation product of heme catabolism, is a non-polar molecule. There are two forms of bilirubin: water-soluble (conjugated or direct) and water-insoluble (unconjugated or indirect) bilirubin. Bilirubin is produced in the endoplasmic reticulum as unconjugated bilirubin, which binds to albumin in plasma and forms albumin-bilirubin complex. This complex is transported to the liver, where it is conjugated with glucuronic acid and forms conjugated bilirubin. Bilirubin has potent antioxidant, anti-inflammatory and autoimmune properties. Bilirubin concentration in human body depends on gender, drug intake, age, etc. Low serum bilirubin is directly correlated with pathological conditions including diabetes mellitus, metabolic syndrome, and cardiovascular diseases. However, high bilirubin indicates hemolysis, jaundice, Gilbert’s syndrome, hepatitis, drug toxicity, and possible blockage of bile ducts. BioVision’s Bilirubin Assay Kit utilizes the Jendrassik-Grof principle to detect bilirubin. Total bilirubin (unconjugated + conjugated) concentration is determined in the presence of a catalyst, where bilirubin reacts with a diazo- salt to form azobilirubin, which absorbs at 600 nm. Direct bilirubin (conjugated) is determined in the absence of catalyst (550 nm).
.