ContactUsLinkComponent
Lowenstein Jensen Medium Base without Starch, Dehydrated Culture Media, HiMedia Laboratories
Lowenstein Jensen Medium Base without Starch, Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95021-266
Supplier:  HiMedia
CAS Number:  
Lowenstein Jensen Medium Base without Starch, Dehydrated Culture Media, HiMedia Laboratories
Catalog # 95021-266
Supplier:  HiMedia
Supplier Number:  M1542-500G
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Lowenstein Jensen Medium Base without Starch
  • Environmentally Preferable:
  • Cat. No.:
    95021-266
  • Supplier no.:
    M1542-500G
Specifications
About this item
Lowenstein Jensen Medium w/o Starch is recommended for susceptibility testing of Mycobacteria with addition of antitubercular drugs. It is recommended for resistance testing by WHO. Lowenstein-Jensen (L-J) Medium without potato starch with drugs incorporated before inspissation is the modification of the International Union Against Tuberculosis (IUAT). Malachite green serves as an inhibitor and also as a pH indicator.Typical Mycobacterium tuberculosis appears granular, rough, warty and dry friable colonies.