Blood Agar Base No.2, Dehydrated Culture Media, HiMedia Laboratories
95022-328
: HiMedia
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Blood Agar Base No. 2
- Environmentally Preferable:
- Cat. No.:95022-328
- Supplier no.:M834-500G
After the addition of blood, the medium permits maximum recovery of streptococci, pneumococci, and other fastidious pathogenic microorganisms without interfering with hemolytic reactions.
- Microbiological culture media
- Packaged in 500 g poly bottles