Order Entry
United States
ContactUsLinkComponent
PCR Decontamination Kit, Enzo Life Sciences
PCR Decontamination Kit, Enzo Life Sciences
Catalog # 76003-302
Supplier:  Enzo Life Sciences
CAS Number:  
PCR Decontamination Kit, Enzo Life Sciences
Catalog # 76003-302
Supplier:  Enzo Life Sciences
Supplier Number:  KIT137-0100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Enzymatic extraction
  • Prep size:
    Mini
  • Refine target molecule:
    ssDNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    PCR Decontamination Kit
  • Size:
    100 Reactions
  • Environmentally Preferable:
  • Description:
    PCR Decontamination Kit
  • Cat. No.:
    76003-302

Specifications

About this item

Rapid removal of contaminating DNA in PCR mastermixes, without reduction in PCR sensitivity.

  • Removes DNA contamination from PCR mastermixes
  • Improves target detection by reducing background
  • Does not effect PCR sensitivity
  • Optimized for PCR and probe based qPCR mixes

For the removal of contaminating DNA from primers and mastermixes prior to PCR.

The PCR decontamination kit uses a dsDNase to remove contaminating DNA from mastermixes. The double-strand specific property allows decontamination in the presence of primers and probe.The dsDNase is irreversibly inactivated by heating to 60°C in the presence of DTT, ensuring that DNA template added to the mastermix after inactivation will not be digested. The mastermix can be used immediately after decontamination to run PCR reactions.

AMPIGENE® Taq polymerases have been treated to remove host DNA and additional clean-up is not necessary. The PCR decontamination kit is recommended for decontamination of primers or competitor’s PCR mixes.

Ordering information: Contains Heat-labile dsDNase, DTT.