Test Tube Brush
Catalog # 470175-994
Supplier: PRECISION MOULDED POLYMERS LIMITED
Specifications
- Bristle material:Synthetic/nylon
- Description:Test tube brush
- Total length:24 cm (9.4")
- Environmentally Preferable:
- Cat. No.:470175-994
- Supplier no.:M76506
Specifications
About this item
Ideal for cleaning test tubes and other tubing apparatus.
Nylon filament mounted in heavy guage twisted wire and completed with double radial tufted end. Synthetic material and nylon bristles provide effective cleaning over the entire bottom of the test tube.
Packaging: Packaged within a heat-sealed polybag.