Order Entry
United States
ContactUsLinkComponent
ZING Green Safety RecycLockout Lockout Tagout, Forklift Propane Tank, ZING Enterprises
ZING Green Safety RecycLockout Lockout Tagout, Forklift Propane Tank, ZING Enterprises
Catalog # 76026-310
Supplier:  ZING Enterprises
   Sustainable Materials
undefined
ZING Green Safety RecycLockout Lockout Tagout, Forklift Propane Tank, ZING Enterprises
Catalog # 76026-310
Supplier:  ZING Enterprises
Supplier Number:  7293
   Sustainable Materials

Specifications

  • Description:
    ZING Forklift Propane Tank Lockout
  • Length:
    3.5"
  • Material:
    Plastic
  • Environmentally Preferable:
    Sustainable Materials
  • Environmental Certification:
    UL Certified
  • Cat. No.:
    76026-310
  • Width:
    3.5"

Specifications

About this item

Hollow cylindrical shell of 100% recycled polycarbonate that surrounds LP gas cylinder handwheels with a padlock (sold separately).

  • Fits electrical LP gas cylinder handwheels up to 2-3/16
  • Made in USA
  • High quality

ZING RecycLockout Forklift Propane Tank Lockout, locks out LP gas cylinder handwheels up to 2-3/16" diameter, accommodates 3 padlocks with shackles up to 9/16" diameter, 100% recycled yellow HDPE with highly visible label "DO NOT OPERATE", UL Validated, supports sustainability initiatives and LEED credits, measures 2-1/2" Dia. x 3-1/2"W x 3-3/8"H, Made in the USA.

Certifications: 29 CFR 1910.147