Order Entry
United States
ContactUsLinkComponent
BD BBL™ Pseudosel Agar Prepared Media Stacker™ Plates, BD Diagnostics
BD BBL™ Pseudosel Agar Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90005-044
Supplier:  BD
CAS Number:  
BD BBL™ Pseudosel Agar Prepared Media Stacker™ Plates, BD Diagnostics
Catalog # 90005-044
Supplier:  BD
Supplier Number:  297882
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Pseudosel Agar (Cetrimide Agar)
  • Environmentally Preferable:
  • Cat. No.:
    90005-044
  • Supplier no.:
    297882

Specifications

About this item

Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.

  • Plates contain a single medium
  • Interlocking, stackable design to minimize sliding or sample disruption
  • Ready for immediate use

Store at 2 to 8 °C (36 to 46 °F).