To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Endothelial Cell
- Clonality:Monoclonal
- Clone:PAL-E
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:1 ml
- Environmentally Preferable:
- Format:PBS buffer with 1% BSA and 0.1% NaN3
- Storage Temperature:Store at 4 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:Endothelial cell antibody was raised in mouse using human melanoma cells as the immunogen.
- Cat. No.:101983-166
- Supplier no.:10RE103A
Specifications
About this item
The endothelium is the thin layer of cells that lines the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. These cells are called endothelial cells. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary. These cells reduce turbulence of the flow of blood, allowing the fluid to be pumped farther.
IHC-F: 1:10
Type: Primary
Antigen: Endothelial Cell
Clonality: Monoclonal
Clone: PAL-E
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human