To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:D-Dimer
- Clonality:Monoclonal
- Clone:M01102704
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Environmentally Preferable:
- Format:Protein A purified and supplied in PBS, pH 7.4, 0.1 % NaN3.
- Storage Temperature:Store at 4 deg C.
- Concentration:Batch dependent - please inquire should you have specific requirements
- Immunogen:D-Dimer antibody was raised in mouse using homogenized fibrin clot D-dimer or high molecular weight fibrin degradation products as the immunogen.
- Cat. No.:101977-740
- Supplier no.:10D42B
Specifications
About this item
D-dimer is a fibrin degradation product, a small protein fragment present in the blood after a blood clot is degraded by fibrinolysis. It is so named because it contains two crosslinked D fragments of the fibrinogen protein. D-dimer concentration may be determined by a blood test to help diagnose thrombosis. Since its introduction in the 1990s, it has become an important test performed in patients suspected of thrombotic disorders.
Optimal conditions to be determined by end user
Type: Primary
Antigen: D-Dimer
Clonality: Monoclonal
Clone: M01102704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human