To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Na+ K+ ATPase alpha 1
- Clonality:Monoclonal
- Clone:C464.6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Format:0.1M Tris Glycine, pH 7.4, with 0.15M NaCl and 0.05% NaN3.
- Storage Temperature:Aliquot and store at -20 deg C. Avoid repeated Freeze/Thaw cycles
- Concentration:Batch dependent - please inquire should you have specific requirements
- Immunogen:Na+ K+ ATPase alpha 1 antibody was raised in mouse using purified Na+/K+ ATPase isolated from membrane fractions of rabbit kidney outer medulla as the immunogen.
- Cat. No.:101983-718
- Supplier no.:10RN102A
Specifications
About this item
Na+/K+-ATPase (also known as the Na+/K+ pump or sodium-potassium pump) is an enzyme located in the plasma membrane (specifically an electrogenic transmembrane ATPase). It is found in the plasma membrane of virtually every human cell and is common to all cellular life. It helps maintain cell potential and regulate cellular volume. In order to maintain the appropriate concentrations, the sodium-potassium pump pumps sodium out and potassium in through active transport.
IHC: 1-10 ug/ml
Type: Primary
Antigen: Na+ K+ ATPase alpha 1
Clonality: Monoclonal
Clone: C464.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rabbit