To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Troponin T (Cardiac)
- Antigen Symbol:TNNT2
- Clonality:Monoclonal
- Clone:M40284
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:1 mg
- Environmentally Preferable:
- Format:Purified by protein A chromatography and supplied in PBS, pH 7.4, with 0.1% NaN3.
- Storage Temperature:Store at 4 deg C.
- Concentration:Batch dependent - please inquire should you have specific requirements
- Immunogen:Troponin T antibody (cardiac) was raised in mouse using free human cTnT as the immunogen.
- Cat. No.:101984-302
- Supplier no.:10T85E
Troponin T is a part of the troponin complex. It binds to tropomyosin, interlocking them to form a troponin-tropomyosin complex. Troponin T binds to tropomyosin and helps position it on actin. It also modulates contraction of striated muscle. The tissue specific subtypes are slow skeletal troponin T1, cardiac troponin T2, and fast skeletal troponin T3.
Optimal conditions to be determined by end user
Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone: M40284
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human