To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Size:2 mg
- Environmentally Preferable:
- Format:Lyophilized from 0.02M K3PO4, pH 7.2, with 0.15M NaCl, 10 mg/ml BSA, and 0.01% NaN3. Immunoglobulin and protease free.
- Storage Temperature:Store at 4 deg C until reconstitution. Following reconstitution aliquot and freeze at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:Goat anti-human IgG (FITC) was raised in goat using human IgG F(c) as the immunogen.
- Cat. No.:102010-132
- Supplier no.:43CCB0958
Specifications
About this item
Immunoglobulin G (IgG) are antibody molecules. Each IgG is composed of four peptide chains - two heavy chains ? and two light chains. Each IgG has two antigen binding sites. Other Immunoglobulins may be described in terms of polymers with the IgG structure considered the monomer.
ELISA: 1:10,000-1:50,000, FC: 1:500-1:2,500, IF: 1:1,000-1:5,000
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human