To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Drebrin
- Clonality:Monoclonal
- Clone:MX823
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:5 ml
- Environmentally Preferable:
- Format:Supplied in liquid form, contains 0.09% NaN3
- Storage Temperature:Store at 2-8 deg C for short term storage. Aliquot and store at -20 deg C for long term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:*Blue Ice*
- Immunogen:Drebrin antibody was raised in mouse using synthetic c-terminal peptide (aa 632-649) coupled to KLH as the immunogen.
- Cat. No.:101981-178
- Supplier no.:10R2460
Specifications
About this item
Drebrins (developmentally regulated brain proteins) are thought to be active in regulating the development and differentiation of neuronal tissue because they are expressed peculiarly in the course of development of neuronal tissues. Drebrin proteins are classified into an embryonic form (E) and an adult form (A) in mammals. The embryonic form is further classified into E1 and E2 in the chicken.
IHC: 1:10 - 1:50, WB: 1:100
Type: Primary
Antigen: Drebrin
Clonality: Monoclonal
Clone: MX823
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: