ContactUsLinkComponent
Eisco® Premium Tabletop Gyroscope
Eisco® Premium Tabletop Gyroscope
Catalog # 470227-638
Supplier:  Wards
undefined
Eisco® Premium Tabletop Gyroscope
Catalog # 470227-638
Supplier:  Wards
Supplier Number:  PH0342
Specifications
  • Description:
    Premium tabletop gyrsocope
  • Environmentally Preferable:
  • Cat. No.:
    470227-638
Specifications
About this item
Beautiful, large, and functional gyroscope for classroom demonstrations.
  • Carefully balanced, adjustable, all-metal construction
  • Fluid motion for seamless performance
  • Permanently balanced on gimbal cradle
Amongst the finest gyroscopes available in the education market, this unit is completely comprised of heavy-duty metals, from gimbals to rotor to base, and has an uncompromisingly fluid motion. A heavy, 7.5 cm brass wheel is perfectly balanced with three pairs of adjustable pivots. The central axis has a small hole in it to wind a string around in order to get the gyroscope spinning. Once spinning, students can push on the different axis to demonstrate how the applied force creates a torque that is perpendicular to the applied force.