About this item
The DR6000™ is the industry's most advanced lab spectrophotometer. It offers high-speed wavelength scanning across the UV and Visible Spectrum, and comes with over 250 pre-programmed methods including the most common testing methods used today. With optional accessories allowing for high-volume testing via a carousel sample changer, and increased accuracy with a sample delivery system that eliminates optical difference errors, this instrument ensures you are ready to handle your wide-ranging water testing needs.
- Optional RFID Technology, for easy method update, sample ID, and certificate of analysis
- Your water testing needs, all in one spectrophotometer
- Accessories for high volume and high accuracy testing needs
- Advanced quality assurance at your fingertips
- Guided procedures and elimination of false readings
- Automatically avoids errors
Your Water Testing Needs, All in One Spectrophotometer—The DR6000 has the most pre-programmed testing methods, including high-speed wavelength scanning across the UV and Visible Spectrum.
Accessories for High Volume and High Accuracy Testing Needs—A carousel sample changer allows up to seven sequential measurements. The Sipper Module, an instrument-controlled sample delivery system, increases precision by constant optical characteristics.
Advanced Quality Assurance at Your Fingertips—The DR6000 comes with integrated QA software for scheduling, documenting and interpreting all of your needed quality measurements.
Guided Procedures and Elimination of False Readings—The DR6000, when used with TNTplus® reagent vials, provides the accurate results you need by guiding you step-by-step through your testing procedures. With TNTplus, the instrument averages 10 readings and eliminates outliers, making scratched, flawed or dirty glassware a non-issue.
Note: RFID technology currently available only in US, Anguilla, American Samoa, Australia, Bolivia, Canada, Cayman Islands, Columbia, Dominican Republic, El Salvador, Federated States of Micronesia, Guam, Guatemala, Marshall Islands, New Zealand, Northern Mariana Islands, Palau, Panama, Puerto Rico, and US Virgin Islands.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
AT-Coagulation, Flocculation and Clarification of Drinking Water AT-Complete Analysis for the Beverage Industry AT-Complete Lab Packages for Brewery Analysis AT-Complete Water Analysis for the Food Industry AT-Determination of High-Range Simplified TKN with TNTplus
Application Notes
AT-DR 6000 Spectrophotometer CSI Specifications AT-DR6000Gäó UV-VIS Spectrophotometer Data Sheet AT-TNT Plus-« Vial Chemistries Data Sheet
Datasheet
FOCUS Industrial Volume 02 Issue 02