About this item
GLP-1 receptor agonist
His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2
Exendin-4 is a 39 aa polypeptide and incretin mimetic which was first isolated from Gila monster saliva. The synthetic form, Exenatide, is a glucagon like peptide-1 (GLP-1) agonist approved for the treatment of diabetes mellitus type II. Research shows Exendin-4 is involved in glucose-dependent enhancement of insulin secretion, reduction of hyperglycemia, glucose-dependent suppression of inappropriately high glucagon secretion, slowing of gastric emptying, and reduction of food intake, often with body weight reduction or blunting of weight gain.
Exendin-4 is a 39 aa polypeptide and incretin mimetic which was first isolated from Gila monster saliva. The synthetic form, Exenatide, is a glucagon like peptide-1 (GLP-1) agonist approved for the treatment of diabetes mellitus type II. Research shows Exendin-4 is involved in glucose-dependent enhancement of insulin secretion, reduction of hyperglycemia, glucose-dependent suppression of inappropriately high glucagon secretion, slowing of gastric emptying, and reduction of food intake, often with body weight reduction or blunting of weight gain.
His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2
Exendin-4 is a 39 aa polypeptide and incretin mimetic which was first isolated from Gila monster saliva. The synthetic form, Exenatide, is a glucagon like peptide-1 (GLP-1) agonist approved for the treatment of diabetes mellitus type II. Research shows Exendin-4 is involved in glucose-dependent enhancement of insulin secretion, reduction of hyperglycemia, glucose-dependent suppression of inappropriately high glucagon secretion, slowing of gastric emptying, and reduction of food intake, often with body weight reduction or blunting of weight gain.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: