To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Fructose-bisphosphate aldolase C
- Antigen Symbol:ALDOC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Environmentally Preferable:
- Epitope:Amino Acid Range = 72-101
- Form:Liquid
- Gene ID:P09972
- Antigen Synonyms:ALDOC
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:39 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This ALDOC antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 72-101 amino acids from the N-terminal region of human ALDOC.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Cat. No.:76010-084
Specifications
About this item
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively.
For WB starting dilution is: 1:1000
Type: Primary
Antigen: ALDOC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human