Order Entry
United States
ContactUsLinkComponent
Medical Chemical (MCC) C and S Transport Medium, Cary Blair Formula
Medical Chemical (MCC) C and S Transport Medium, Cary Blair Formula
Catalog # 75993-506
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Medical Chemical (MCC) C and S Transport Medium, Cary Blair Formula
Catalog # 75993-506
Supplier:  Hardy Diagnostics
Supplier Number:  280505
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Culture and sensitivity transport medium, Cary Blair formula
  • Size:
    100/case
  • Environmentally Preferable:
  • Storage Temperature:
    15...25 °C
  • Cat. No.:
    75993-506
  • Shelf life:
    1800
  • Application:
    Clinical

Specifications

About this item

Culture and sensitivity medium is recommended for the recovery of Shigella, Salmonella, Campylobacter and Vibrio species.

  • Includes pH indicator
  • Aqueous solution

A pH indicator is included to detect acidic conditions which are harmful for the recovery of enteric pathogens. An aqueous solution of sodium thioglycolate, agar, and inorganic salts.