Homofermentative-Heterofermentative Differential (HHD) Medium, Hardy Diagnostics
Catalog # 75994-472
Supplier: Avantor
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Homofermentative-Heterofermentative Differential (HHD) Medium
- Size:15 x 100 mm
- Environmentally Preferable:
- Minimum Order Quantity:10 packs
- Cat. No.:75994-472
Specifications
About this item
Use in the enumeration and differentiation of homofermentative and heterofermentative lactic acid bacteria.