ALLWIK® Universal Absorbent Pillows, Brady® SPC® Absorbents
Catalog # 10810-750
Supplier: Brady Worldwide
Specifications
- Color:Gray
- Description:ALLWIK® Universal Absorbent Pillows
- Length:18"
- Material:PP
- Environmentally Preferable:
- Cat. No.:10810-750
- Width:18"
- Supplier no.:AW1818
Specifications
About this item
Use ALLWIK® all-purpose absorbents to clean up oils, coolants, solvents, and water-based fluids throughout any facility.
- Designed for use in tight spaces, under valves, or under machinery lifted off the floor
- Pillows provide the greatest absorbency per square inch
- Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
- Gray color masks spills and stains, helping users resist the urge to pick up the absorbent before it is fully saturated
- Made of all-purpose polypropylene suitable for most industrial applications
ALLWIK® Pillows feature a dark spunbond outer covering filled with gray polypropylene. Versatile enough for use with all oil and water-based fluids.
Certifications: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).