Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
GREENCAST®-94 GR PLUS
GREENCAST®-94 GR PLUS 94% Alumina
GREENCAST®-94 GR PLUS 94% Alumina
GREENCAST®-94 GR PLUS 94% Alumina
Catalog # 11019-250
Supplier:
HarbisonWalker
CAS Number:
Expand image
GREENCAST®-94 GR PLUS 94% Alumina
Catalog # 11019-250
Supplier:
HarbisonWalker
Supplier Number:
59322
CAS Number:
Specifications
Size:
55 lbs.
Environmentally Preferable:
Cat. No.:
11019-250
Supplier no.:
59322
Specifications
About this item
Product Details
:
CAS Number:
65997-16-2
Product Details