Order Entry
United States
ContactUsLinkComponent
Human Recombinant Insulin, MP Biomedicals
Human Recombinant Insulin, MP Biomedicals
Catalog # IC0219390010
Supplier:  MP Biomedicals
CAS Number:  
Human Recombinant Insulin, MP Biomedicals
Catalog # IC0219390010
Supplier:  MP Biomedicals
Supplier Number:  0219390010
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Bottle
  • CAS No.:
    11061-68-0
  • Protein/Peptide Type:
    Recombinant
  • Source:
  • Species:
    Human
  • Size:
    10 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • MDL No.:
    MFCD00131380
  • Protein/Peptide Name:
    Insulin
  • Solubility:
    Insulin is not soluble at neutral pH. It can be solubilized (1 - 10 mg/mL) in dilute acetic acid or dilute hydrochloric acid, pH 2-3.
  • Cat. No.:
    IC0219390010

Specifications

About this item

Methods for immobilizing insulin on polystyrene dishes have been reported for applications involving protein-free cell cultures.

Typical Working Concentration: 1 to 10 ug/mL for cell culture. Unit definition: 1 unit corresponds to the efficiency of 0.04167 mg international standard substance. An HPLC method exists to test the potency.