About this item
Tergitol™ NP-7, Surfactant is a Nonylphenol Ethoxylate surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.
- Excellent detergency
- Outstanding wetting
- Versatile solubility characteristics
- Low odor
- Soluble in water
Density at 20°C (68°F): 1.044 g/ml
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.