Tergitol™ 15-S-3 is a Dow Chemical secondary alcohol ethoxylate, non-ionic surfactant used in a wide variety of applications including pre-wash spot removers, defoamers, textiles, pulp and paper and dry cleaners.
- Non-ionic surfactant
- Excellent hydrocarbon compatibility and detergency
- Excellent oil soluble emulsifier
- Superior handling properties
- Insoluble in water
Density at 20°C (68°F): 0.918 g/ml