Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Supplier: SI Analytics
Certificates
About this item
Suitable for cleaning combination electrodes and reference electrodes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.