ContactUsLinkComponent
Orion™ Star™ Series RS232 to USB Cable Adapter, Thermo Scientific
Orion™ Star™ Series RS232 to USB Cable Adapter, Thermo Scientific
Catalog # 10010-666
Orion™ Star™ Series RS232 to USB Cable Adapter, Thermo Scientific
Catalog # 10010-666
Supplier Number:  1010005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    RS232 to USB cable adapter
  • Environmentally Preferable:
  • Cat. No.:
    10010-666
  • Supplier no.:
    1010005
Specifications
About this item
This cable adapter can be used to connect a USB port to the RS232 port of a meter.
The adapter allows interface between a USB connection and Orion™ Versa Star™ meters, Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters, and legacy Orion™ Star™ and Star Plus meters.