Differential Quik Stain Kit (Modified Giemsa) is a high quality, rapid turnaround staining kit that serves many purposes in the cost conscious laboratory environment.
- Rapid stain for immediate interpretation of fine needle aspiration biopsies
- Differential stain designed to show virtually all dermapathology cells
- Visualize nuclear and cytoplasmic staining easily
Differential Quik Stain Kit may be used for rapid blood smears for differential assessment, as well as detecting H. pylori microorganisms. The stain is very useful to pathologists for immediate interpretation of fine needle aspiration biopsies. Kit contains a fixative for air-dried cell suspensions and/or touch preparation slides.