ContactUsLinkComponent
Mini Tischler Micro-Cut Biopsy Punch, OR Grade, Sklar
Mini Tischler Micro-Cut Biopsy Punch, OR Grade, Sklar
  10194-152
 :  Sklar
undefined
Mini Tischler Micro-Cut Biopsy Punch, OR Grade, Sklar
  10194-152
 :  Sklar
 :  90-8945
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Micro-Cut Mini-Tischler Biopsy Punch
  • Length:
    20.6 cm (8")
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Tip configuration:
    Semi Elliptical/Semi Oval Bite with 1 Lower Tooth
  • Curvature:
    Straight
  • Cat. No.:
    10194-152
  • Shape:
    Rectangular
  • Supplier no.:
    90-8945
  • Tip type:
    With locking tip
Sklar's Micro-Cut Mini Tischler Biopsy Punch Forceps are a non-ratcheted, double spring instrument used to cut tissue samples in gynecologic procedures.
This product is straight with an 8 inch shaft and 2.3mm x 4.2mm rectangular bite. It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers. Micro-Cut biopsy punches are identified by gold handles.
 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.