Order Entry
United States
ContactUsLinkComponent
TBARS Assay Kit, Cayman Chemical
TBARS Assay Kit, Cayman Chemical
Catalog # 75816-718
CAS Number:  
undefined
TBARS Assay Kit, Cayman Chemical
Catalog # 75816-718
Supplier Number:  10009055-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:
  • Cat. No.:
    75816-718
  • Range:
    0.0625-50 µM
  • Includes:
    Kit contains: Standard, Reagents, and Plates
  • Supplier no.:
    10009055-96

Specifications

About this item

For the measurement of lipid peroxidation in plasma, serum, urine, tissue homogenates, and cell lysates.

  • Measure TBARS in plasma, serum, urine, tissue homogenates, and cell lysates
  • Assay 40 samples in duplicate
  • Assay range: 0.0625-50 µM
  • Plate-based colorimetric measurement (530-540 nm) or fluorometric measurement (ex 530 nm, em 550 nm)

Oxidative stress in the cellular environment results in the formation of highly reactive and unstable lipid hydroperoxides. Decomposition of the unstable peroxides derived from PUFAs results in the formation of malondialdehyde (MDA), which can be quantified colorimetrically following its controlled reaction with thiobarbituric acid. The measurement of these 'Thiobarbituric Acid Reactive Substances' (TBARS) is a well-established method for screening and monitoring lipid peroxidation. Modifications of the TBARS assay has been used to evaluate a wide range of samples that include human and animal tissues and fluids, drugs and foods. Even though there remains a controversy cited in literature regarding the specificity of TBARS toward compounds other than MDA, it still remains the most widely employed assay used to determine lipid peroxidation. Cayman’s TBARS Assay Kit provides a simple, reproducible, and standardized tool for assaying lipid peroxidation in plasma, serum, urine, tissue homogenates, and cell lysates.