About this item
Fungus, GMS, Multi-tissue, artificial control slides contain sections of Aspergillus sp. positive staining rat lung, Candida sp. positive staining rat lung, Histoplasma sp. positive staining animal tissue, and negative staining human lung.
- Tissue Type: (+) Aspergillus Rat Lung / (+) Candida Rat Lung / (+) Histoplasma Animal Tissue / (-) Lung
- All control tissue is sectioned on Superfrost® Plus slides
- Staining validation technical memo included in each set
- All tissues are formalin fixed, paraffin embedded FFPE
- All slides are air dried
- Histo slides are cut at 4 µm, unless otherwise specified
- Standard tissue placement is mid-high on slide
Newcomer supply fungus, GMS, multi-tissue, artificial control slides, use a variety of lung tissue sources for the positive histochemical staining of Aspergillus sp., Candida sp., and organisms exhibiting morphology consistent with Histoplasma sp
Staining Results:
Aspergillus sp. will stain sharply outlined in black.
Candida sp. will stain sharply outlined in black.
Histoplasma sp. will stain sharply outlined in black.
Background will stain green.
Negative lung will stain negative for fungus.
These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.
in separate tissue sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.