Specifications
- Application:
- Description:MyTaq™ HS Red Mix
- Size:1000 reactions
- Environmentally Preferable:
- Cat. No.:490005-938
- Volume:20× 1.25 ml
- Supplier no.:BIO-25048
Specifications
About this item
MyTaq™ HS Red Mix is high-performance ready-to-use, direct gel load, red colored, 2x mix for fast, highly-specific hot-start PCR
- Universal DNA polymerase - Complete PCR reaction in less than 30 minutes
- Antibody-based hot start polymerase
- Highest specificity and superior performance
- All-in-one mastermix formulations enhance reproducibility
They have wide range of application in Fast PCR reactions, Colony PCR, Genotyping, Multiplexing, Low-copy PCR assays, Specific amplification of difficult templates (GC-rich) and Assays with prolonged reaction set-up etc.
It is powered by antibody mediated hot-start and does not possess polymerase activity during the reaction set-up, thus reducing non-specific amplification. The advanced formulation of MyTaq™ HS Mix allows fast cycling conditions to be used, greatly reducing the reaction time without compromising PCR specificity and yield.