Order Entry
United States
ContactUsLinkComponent
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # BJ31087-25L
CAS Number:  
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # BJ31087-25L
Supplier Number:  31087-25L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    25 L
  • Environmentally Preferable:
  • Cat. No.:
    BJ31087-25L
  • Supplier no.:
    31087-25L

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
 :
Formula:HCl
MDL Number:MFCD00011324
 :36.46
UN:1789
 :GHS07
 :GHS05
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8 III