To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD10 molecule
- Antigen Symbol:CD10
- Clonality:Monoclonal
- Clone:CB-CALLA
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:4311
- Antigen Synonyms:NEP|gp100|CALLA|common acute lymphoblastic leukemia antigen|KII-NA|MME|enkephalinase
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Human CALLA cells
- Cat. No.:101442-950
- Supplier no.:9541-09
Specifications
About this item
CD10, also known as the common acute lymphoblastic leukemia antigen (CALLA), is a type II transmembrane glycoprotein. It is expressed on granulocytes, a subset of B progenitor cells, germinal center cells, and fibroblasts. CD10 is a zinc-binding metalloprotease which is thought to down-regulate cellular responses to peptide hormones.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: CB-CALLA
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human