To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD28
- Antigen Symbol:CD28
- Clonality:Monoclonal
- Clone:PV-1
- Flow Cytometry:Yes
- Host:Hamster
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Mouse
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:12487
- Antigen Synonyms:Tp44|T44
- Storage Buffer:Supplied in PBS
- Storage Temperature:–20 °C
- Concentration:0.5 mg/mL
- Shipping Temperature:Dry Ice
- Immunogen:C57BL/6N mouse T cell lymphoma EL4 cell line
- Tested Applications:Costim, Stim
- Cat. No.:100243-400
- Supplier no.:1615-14
Specifications
About this item
CD28 is a type I disulfide-linked homodimer that is constitutively expressed on most thymocytes, at low density on nearly all CD4+ and CD8+ peripheral T lymphocytes and at very low levels on NK cells. Its expression is upregulated upon T-cell activation. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) on B cells and other antigen presenting cells and plays an important role in the interaction between T cells and B cells. CD28 is a costimulatory receptor involved in many, but not all, T-cell independent immune responses.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: PV-1
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse