To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G4 F(c)
- Antigen Symbol:IgG4 F(c)
- Clonality:Monoclonal
- Clone:HP6025
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3503
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Human IgG4 myeloma protein
- Tested Applications:FLISA, Multiplex
- Cat. No.:100245-818
- Supplier no.:9200-09
Specifications
About this item
PE Anti-Human IgG4 Fc antibody for use in flow cytometry and multiplex assays.
Type: Secondary
Antigen: IgG4 F(c)
Clonality: Monoclonal
Clone: HP6025
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human