To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Immunoglobulin Y
- Antigen Symbol:IgY
- Clonality:Monoclonal
- Clone:G-1
- Conjugation:Biotin
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Turkey,Chicken
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Affinity purified chicken Ig or isolated lymphocytes
- Cat. No.:100245-522
- Supplier no.:8320-08
Specifications
About this item
Biotin Anti-Chicken IgY antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, ELISA, and immunoprecipitation assays.
Type: Primary
Antigen: IgY
Clonality: Monoclonal
Clone: G-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Turkey