To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD3
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:CT-3
- Conjugation:Spectral Red® RPE (R-Phycoerythrin)/Cy5®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Pigeon
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:396518
- Antigen Synonyms:T3/TCR complex
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.1 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Chicken thymocytes and Ig- blood mononuclear cells
- Cat. No.:100245-416
- Supplier no.:8200-13
Specifications
About this item
CD3 is a member of the T cell receptor-associated CD3 complex. The monoclonal antibody CT-3 recognizes a complex of at least three polypeptides of Mr 20, 19, and 17 kDa (two of which are N-glycosylated) on chicken T cells. The antibody also coprecipitates a polypeptide of 90 kDa from digitonin solubilized T cell lysates, which can be reduced to two polypeptides of Mr 50 and 40 kDa.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: CT-3
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Pigeon