Streptavidin MagneSphere Paramagnetic Particles, Promega
Supplier: Promega Corporation
Certificates
About this item
Streptavidin MagneSphere Paramagnetic Particles (PMPs) are used for magnetic separation and purification of a variety of biotinylated nucleic acid or protein molecules.
- Separation and Purification of Biotinylated Nucleic Acids or Proteins
- Strong, specific binding to biotinylated molecules
- Enable binding, washing and magnetic separation from undesired materials
- Used for purification of DNA, mRNA and proteins.
- Supplied at a concentration of 1mg/ml
The Streptavidin MagneSphere Paramagnetic Particles (PMPs) may be used in the magnetic separation and purification of a wide variety of biotinylated nucleic acid or protein molecules. The particles are quality-tested and approved for isolation of biotinylated nucleic acids, proteins and antibodies.
They feature strong, specific binding to biotinylated molecules.
Specifications
- Catalog No:
- PAZ5482
- PAZ5481
- Storage Conditions:
- Store at 4 °C (2 to 10 °C). Do not freeze the paramagnetic particles.
- Store at 4 °C (2 to 10 °C). Do not freeze the paramagnetic particles.