About this item
HeLaScribe Nuclear Extract, in vitro Transcription Grade.
- Cell-Free Extract for in vitro Transcription of Eukaryotic Genes
- Component of the HeLaScribe(R) Nuclear Extract in vitro Transcription System
- Source of transcription factors, DNA-binding proteins and RNA processing components
HeLaScribe Nuclear Extract, in vitro Transcription Grade, derived from HeLa cell nuclei, provides a cell-free system for in vitro transcription of eukaryotic genes.
The HeLa Nuclear Extract included in the HeLaScribe Nuclear Extract In-Vitro Transcription System is prepared by a modification of the method of Dignam et al. Extracts prepared by this method have been shown to allow transcription from the human transferrin gene promoter and the adenovirus two major late promoter. The system also includes all of the necessary components for in-vitro transcription as well as a positive control template (CMV immediate early promoter DNA).
The nuclear extract is also a source for a variety of transcription factors, DNA-binding proteins, and the enzymatic machinery involved in RNA processing.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: