Specifications
- Application:
- Description:T7 RiboMAX Express Large Scale RNA Production System, 1 system
- Size:1 system
- Environmentally Preferable:
- Cat. No.:PAP1320
- Supplier no.:P1320
Specifications
About this item
The T7 RiboMAX Express Large Scale RNA Production System is an in vitro transcription system designed to produce milligram amounts of RNA in a short time.
- Produces 5-8.5mg/ml RNA in 30 Minutes
- Consistent production of milligram amounts of RNA
- Minimal pipetting steps and setup time
- Efficiently transcribes DNA templates of varying sizes. Works with transcripts as short as 21bp.
The T7 RiboMAX Express Large Scale RNA Production System is an in vitro transcription system designed for the consistent production of milligram amounts of RNA in a short amount of time. Due to optimization of the enzyme mix and transcription buffer, yields of 5-8.5mg/ml are generated in 30 minutes, compared to 2-4 hours with other commercially available systems. To minimize pipetting steps and errors, the 2X transcription buffer includes all four rNTPs. In addition, the system includes RQ1 RNase-Free DNase for the removal of plasmid template after transcription. Due to the combined 2X buffer and rNTPs, the T7 RiboMAX Express System is not recommended for the synthesis of RNA for applications that require capped RNA. For synthesis of capped RNA, please order the standard RiboMAX Large Scale RNA Production System---T7 (Cat.# P1300).
Due to the combined 2X buffer and rNTPs, the T7 RiboMAX™ Express System is not recommended for the synthesis of capped RNA. For synthesis of capped RNA, please order the standard RiboMAX Large-Scale RNA Production System.
To minimize pipetting steps and errors, the 2X transcription buffer includes all four rNTPs. The system also includes RQ1 RNase-Free DNase for the removal of plasmid template after transcription.