Order Entry
United States
ContactUsLinkComponent
pGEM-3Zf(+) Vector, 20 µg, Promega
pGEM-3Zf(+) Vector, 20 µg, Promega
Catalog # PAP2271
CAS Number:  
undefined
pGEM-3Zf(+) Vector, 20 µg, Promega
Catalog # PAP2271
Supplier Number:  P2271
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    pGEM®-3Zf(+) Vector
  • Size:
    20 μg
  • Environmentally Preferable:
  • Storage Conditions:
    Store vector at –20°C and bacterial strain at –70°C
  • Cat. No.:
    PAP2271
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    P2271

Specifications

About this item

Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.

  • Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
  • Blue/white screening for easy identification of recombinant clones
  • In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • The difference between + and - vectors is orientation of the f1 origin

The pGEM-3Zf(+) and pGEM-3Zf(-) Vectors are derived from the pGEM-3Z Vector and contain the origin of replication of the filamentous phage f1. These plasmids contain T7 and SP6 RNA polymerase promoters flanking a multiple cloning region within the alpha-peptide coding region of beta-galactosidase. Insertional inactivation of the alpha-peptide allows recombinant clones to be identified directly by color screening on indicator plates when using appropriate E. coli strains (e.g., JM109). The multiple cloning region contains unique restriction sites for EcoRI, SacI, KpnI, AvaI, SmaI, BamHI, XbaI, SalI, AccI, HincII, PstI, SphI and HindIII. The pGEM-3Zf(+) and -3Zf(-) Vectors are identical except for the orientation of the f1 origin and can be used as standard cloning vectors, as templates for in vitro transcription and for the production of circular ssDNA.

The multiple cloning region contains unique restriction sites for EcoRI, SacI, KpnI, AvaI, SmaI, BamHI, XbaI, SalI, AccI, HincII, PstI, SphI, and HindIII. The pGEM-3Zf(+) and -3Zf(–) Vectors are identical except for the orientation of the f1 origin and can be used as standard cloning vectors, as templates for in-vitro transcription, and for the production of circular ssDNA.

Insertional inactivation of the α-peptide allows recombinant clones to be identified directly by color screening on indicator plates when using appropriate E. coli strains (e.g., JM109).