ContactUsLinkComponent
Test Tube Brush, Chemglass
  10034-292
 :  Chemglass
Small Business Enterprise
undefined
Test Tube Brush, Chemglass
  10034-292
 :  Chemglass
 :  CG-1143-01
Small Business Enterprise
  • Bristle color:
    White
  • Bristle material:
    PP
  • Handle material:
    Plastic
  • Description:
    Test Tube Brush
  • Environmentally Preferable:
  • Cat. No.:
    10034-292
  • Handle color:
    White
  • Supplier no.:
    CG-1143-01
This brush has polypropylene bristles and can be spread apart to clean tubes having an I.D. of 14 mm or greater.