Order Entry
United States
ContactUsLinkComponent
Specimen Bags, Biohazard, Platinum code, Platinum Plus
Specimen Bags, Biohazard, Platinum code, Platinum Plus
Catalog # 10852-790
Supplier:  CARESFIELD, LLC
undefined
Specimen Bags, Biohazard, Platinum code, Platinum Plus
Catalog # 10852-790
Supplier:  CARESFIELD, LLC
Supplier Number:  48609SB
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Specimen bag, biohazard, back pouch and zip-top
  • Dimensions:
    15.2×22.8 cm (6×9")
  • Environmentally Preferable:
  • Cat. No.:
    10852-790
  • Supplier no.:
    48609SB

Specifications

About this item

These specimens bags are user friendly and provides a safe and compliant method of transporting specimens.

  • A separate back pouch is included on the back for storage of documents
  • Moisture-resistant bags protect specimen from environmental effects
  • Absorbent sheets are available in bags to soak up moisture

Zip-top for easy and secure closure, and an optional tear-off top for fast specimen access.

Packaging: 1000 bags per case (10 dispenser bags of 100)