ContactUsLinkComponent
HIGHDEF® DAB Chromogen/Substrate Set, Enzo Life Sciences
HIGHDEF® DAB Chromogen/Substrate Set, Enzo Life Sciences
Catalog # 75841-138
Supplier:  Enzo Life Sciences
CAS Number:  
HIGHDEF® DAB Chromogen/Substrate Set, Enzo Life Sciences
Catalog # 75841-138
Supplier:  Enzo Life Sciences
Supplier Number:  ACC105-0200
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    HIGHDEF® DAB Chromogen/Substrate Set
  • Environmentally Preferable:
  • Cat. No.:
    75841-138
  • Volume:
    200 ml
  • Supplier no.:
    ACC105-0200
Specifications
About this item
Used in conjunction with peroxidase-based immunostaining systems.
  • Peroxidase reacts with hydrogen peroxide substrate to degrade it
  • Use with peroxidase-based immunostaining systems
The HIGHDEF® DAB Chromogen/Substrate Set can be used in conjunction with peroxidase-based immunostaining systems. Peroxidase from the antibody detection system reacts with hydrogen peroxide substrate to degrade it, which then reacts with DAB, precipitating it at positive sites and yielding a dark brown color.
Ordering information: Contains HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate Buffer.