Sodium chloride USP
Catalog # 10142-842
Supplier: Hawkins Pharmaceutical Group
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:50 kg
- Environmentally Preferable:
- Cat. No.:10142-842
- Supplier no.:14734
Specifications
Specification Test Results
- Identification A (Na):Conforms
- Identification B (Cl):Conforms
- Aluminium (Al):≤0.2 ppm
- Arsenic (As):≤1 ppm
- Barium (Ba):Conforms
- Ferrocyanides:Conforms
- Iodidies (I):Conforms
- Iron (Fe):≤2 ppm
- Limit of Bromides:≤100 ppm
- Limit of Phosphates:≤25 ppm
- Limit of Potassium:≤500 ppm
- Magnesium and Alkali Earth Metals:≤100 ppm
- Nitrites (NO₂) @ 354 nm:≤0.01
- Sulfate (SO₄):≤200 ppm
- Heavy Metals:≤5 ppm
- Appearance of solution:Clear and Colorless
- Loss on Drying @105 °C:≤0.5%
- Acidity or Alkalinity:Conforms
- Bacterial Endotoxins<85>:≤5 IU/g (per EP)
- Assay:99.0 - 100.5%
Specification Test Results
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Boiling Pt:1413
Melting Pt:801
Merck Index:14,08599