Mini Vortex Mixer, Greiner Bio-One
Catalog # 10818-595
Supplier: Greiner Bio-One
Specifications
- Description:Mini Vortex Mixer
- Environmentally Preferable:
- Cat. No.:10818-595
- Speed range:2800 rpm (fixed)
Specifications
About this item
Compact, powerful, vortex mixer designed for vortexing tubes of various sizes (0.4 to 50 mL).
- Touch mix mode
- Fixed speed
Rubber cup head is standard for single tube mixing. The mini vortex mixer is intended only for indoor use predominantly in research laboratories.