Order Entry
United States
ContactUsLinkComponent
Anti-Strep-tag II Rabbit Polyclonal Antibody
Anti-Strep-tag II Rabbit Polyclonal Antibody
Catalog # 10178-558
Supplier:  Avantor
Anti-Strep-tag II Rabbit Polyclonal Antibody
Catalog # 10178-558
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Strep-tag II
  • Antigen Symbol:
    Strep-tag II
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen Synonyms:
    StrepII-tag
  • Storage Buffer:
    Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage Temperature:
    Antibody can be stored at 4 °C for three months and at –20 °C for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/ml
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Strep-tag II antibody was raised in rabbits against the amino acid sequence ”srWSHPQFEKsh”.
  • Purification:
    Affinity chromatography Purified via peptide column.
  • Cat. No.:
    10178-558
  • Supplier no.:
    PK-AB718-4335

Specifications

About this item

Rabbit Anti-Strep-tag II (CT)

StrepII-tag antibody can be used for the detection of StrepII by Western blot at 1 µg/mL.

Type: Primary
Antigen: Strep-tag II
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Strep-Tag Ii