To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Apoptosis Antigen 1/Fas
- Antigen Symbol:APO-1/Fas (APO-1-1)
- Clonality:Monoclonal
- Clone:APO-1-1
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:Fas
- Storage Buffer:PBS
- Storage Temperature:Stable at 2-8°C. After opening, aliquote contents and freeze at –20 °C for long-term storage. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:2-8°C
- Purification:> 95% IgG1 as determined by SDS-gel electrophoresis.
- Cat. No.:10175-054
- Supplier no.:PK-AB704-69208
Specifications
About this item
Mouse Anti-Human Apoptosis Antigen 1/Fas (clone APO-1-1)
Specifically recognizes human APO-1.
Type: Primary
Antigen: APO-1/Fas (APO-1-1)
Clonality: Monoclonal
Clone: APO-1-1
Conjugation:
Epitope:
Host: Mouse
Isotype: mouse IgG1
Reactivity: Human